Antimony(III) sulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Diantimony tris(sulphate)
10783-50G 10783-10G
SIAL10783-50GEA 464 EUR
SIAL10783-50G SIAL10783-10G
Antimony(III) sulphate, Sigma-Aldrich®
Antimony (III) sulphate
Formula: O₁₂S₃Sb₂
MW: 531.71 g/mol
Density: 3.6246 g/cm³ (20 °C)
CAS Number: 7446-32-4
EINECS: 231-207-6
UN: 3077
ADR: 9 III
Merck Index: 12,00745