Dimethyl methylphosphonate, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
D169102-100G
D169102-500G
D169102-5G
SIALD169102-100GEA
53.3
EUR
SIALD169102-100G
SIALD169102-500G
SIALD169102-5G
Dimethyl methylphosphonate, Sigma-Aldrich®
Dimethyl methylphosphonate
Formula:
C₃H₉O₃P MW: 124.08 g/mol Boiling Pt: 181 Density: 1.1596 g/cm³ (20 °C) |
MDL Number:
MFCD00008349 CAS Number: 756-79-6 EINECS: 212-052-3 Merck Index: 13,03425 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...