Tributylphosphine, Sigma-Aldrich®

Supplier: Merck

Synonyms: Tri-n-butylphosphine, TBP

M-Clarity™ quality level = MQ200

T49484-25ML T49484-5ML
SIALT49484-25MLEA 195 EUR
SIALT49484-25ML SIALT49484-5ML
Tributylphosphine, Sigma-Aldrich®
Tributylphosphine
Formula: C₁₂H₂₇P
MW: 202.32 g/mol
Boiling Pt: 240
Melting Pt: –65 °C
Density: 0.8165 g/cm³ (20 °C)
Flash Pt: 40
MDL Number: MFCD00009462
CAS Number: 998-40-3
EINECS: 213-651-2
UN: 3254
ADR: 4.2 I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR