Cetrimonium bromide

Supplier: G Bioscienes

DG094EA 94 EUR
DG094
Cetrimonium bromide
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364,45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
MDL Number: MFCD00011772
CAS Number: 57-09-0
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR