Tetrabutylammonium bromide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
193119-100G
193119-25G
193119-500G
193119-5KG
SIAL193119-100GEA
72.5
EUR
SIAL193119-100G
SIAL193119-25G
SIAL193119-500G
SIAL193119-5KG
Tetrabutylammonium bromide, Sigma-Aldrich®
Tetrabutylammonium bromide
Formula:
C₁₆H₃₆BrN Melting Pt: 100…103 °C Density: 1.148 g/cm³ (20 °C) |
MDL Number:
MFCD00011633 CAS Number: 1643-19-2 EINECS: 216-699-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...