Tetramethylammonium chloride
Supplier: Roth Carl
Danger
4747.1 4747.2
ROTH4747.1EA 0 EUR
ROTH4747.1 ROTH4747.2
Tetramethylammonium chloride
Tetramethylammonium chloride
Formula: (CH₃)₄NCl
Melting Pt: > 300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III