Tetramethylammonium chloride, Sigma-Aldrich®
Supplier: Merck
Danger
T19526-100G T19526-500G T19526-5G
SIALT19526-100GEA 42.2 EUR
SIALT19526-100G SIALT19526-500G SIALT19526-5G
Tetramethylammonium chloride, Sigma-Aldrich®
Tetramethylammonium chloride
Formula: (CH₃)₄NCl
Melting Pt: > 300 °C
Density: 1.169 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00011628
CAS Number: 75-57-0
EINECS: 200-880-8
UN: 2811
ADR: 6.1,III