Acetylcholine chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2-Acetoxyethyl)trimethylammonium chloride, 2-(Acetyloxy)-N,N,N-trimethylethanaminium chloride, 2-Acetoxy-N,N,N-trimethylethanaminium chloride
M-Clarity™ quality level = MQ400
A2661-100G A2661-25G
SIALA2661-100GEA 272 EUR
SIALA2661-100G SIALA2661-25G
Acetylcholine chloride, Sigma-Aldrich®
Acetylcholine chloride
Formula: H₃CC(O)O(CH₂)₂N(CH₃)₃Cl
Melting Pt: 151 °C
Density: 1.263 g/cm³ (20 °C)
MDL Number: MFCD00011698
CAS Number: 60-31-1
EINECS: 200-468-8
Merck Index: 13,00089