Tetraethylammonium iodide 98
Supplier: Thermo Fisher Scientific
Warning
Synonyms: TEAI, Tetrylammonium iodide
A11783.0B A11783.18 A11783.30
A11783.0BEA 507 EUR
A11783.0B A11783.18 A11783.30
Tetraethylammonium iodide 98
Tetraethylammonium iodide
Purity: 98+% CAS No.: 68-05-3 Molecular Formula: (CH3CH2)4NI Formula Weight: 257.17 Melting Point: ca 320° dec. Beilstein Reference: 3562649
Formula: C₈H₂₀IN
MW: 257.16 g/mol
Melting Pt: ∼ 320 °C (dec.)
Density: 1.566 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011829
CAS Number: 68-05-3
EINECS: 200-676-9