Butyrylthiocholine iodide, Sigma-Aldrich®

Supplier: Merck

Synonyms: S-n-Butyrylthiocholine iodide, (2-Butyrylthioethyl)trimethylammonium iodide, 2-(Butyrylthio)-N,N,N-trimethylethan-1-aminium iodide, (Propylcarbonylthioethyl)trimethylammonium iodide, (2-(Butyrylthio)ethyl)trimethylammonium iodide

B3253-25G B3253-5G
SIALB3253-25GEA 147 EUR
SIALB3253-25G SIALB3253-5G
Butyrylthiocholine iodide, Sigma-Aldrich®
Butyrylthiocholine iodide
Formula: H₃C(CH₂)₂C(S)O(CH₂)₂N(CH₃)₃I
MW: 317.23 g/mol
Melting Pt: 172 °C
Storage Temperature: Freezer
MDL Number: MFCD00011845
CAS Number: 1866-16-6
EINECS: 217-475-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR