Tetrabutylammonium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
M-Clarity™ quality level = MQ100
155837-100G 155837-1KG 155837-25G
SIAL155837-100GEA 109 EUR
SIAL155837-100G SIAL155837-1KG SIAL155837-25G
Tetrabutylammonium hydrogen sulphate, Sigma-Aldrich®
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
MW: 339.54 g/mol
Melting Pt: 169…172 °C
Density: 1.01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5