Tetrabutylammonium hydrogen sulphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetrabutylammonium bisulphate, Tetrabutylammonium hydrogensulphate
86868-100G 86868-25G 86868-500G
SIAL86868-100GEA 234 EUR
SIAL86868-100G SIAL86868-25G SIAL86868-500G
Tetrabutylammonium hydrogen sulphate, Sigma-Aldrich®
Tetrabutylammonium hydrogen sulphate
Formula: (C₄H₉)₄NHSO₄
Melting Pt: 169…172 °C
Density: 1.01 g/cm³ (25 °C)
MDL Number: MFCD00011637
CAS Number: 32503-27-8
EINECS: 251-068-5