Benzethonium chloride, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Hyamine
SIAL53751-250GEA
471
EUR
SIAL53751-250G
SIAL53751-50G
Benzethonium chloride, Sigma-Aldrich®
Benzethonium chloride
Formula:
C₂₇H₄₂ClNO₂ MW: 448,09 g/mol Melting Pt: 164…166 °C |
MDL Number:
MFCD00011742 CAS Number: 121-54-0 UN: 3077 ADR: 9,III Merck Index: 13,01074 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...