Tris(2-methylphenyl)phosphine, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tris(o-tolyl)phosphine, Tri-o-tolylphosphine
M-Clarity™ quality level = MQ200
287822-10G 287822-1G
SIAL287822-10GEA 354 EUR
SIAL287822-10G SIAL287822-1G
Tris(2-methylphenyl)phosphine, Sigma-Aldrich®
Tris(2-methylphenyl)phosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 187…192 °C (0.05 mmHg)
Melting Pt: 125.5 °C
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9