Triphenylantimony, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Triphenylstibine
M-Clarity™ quality level = MQ200
T81809-100G
T81809-25G
SIALT81809-100GEA
80.4
EUR
SIALT81809-100G
SIALT81809-25G
Triphenylantimony, Sigma-Aldrich®
Triphenylantimony
Formula:
C₁₈H₁₅Sb MW: 353.08 g/mol Boiling Pt: 377 °C (1013 hPa) Melting Pt: 52…54 °C Density: 1.5 g/cm³ (12 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003000 CAS Number: 603-36-1 EINECS: 210-037-6 UN: 2811 ADR: 6.1,III |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...