Tetraphenylphosphonium chloride, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
218790-10G 218790-50G 218790-5G
SIAL218790-10GEA 117 EUR
SIAL218790-10G SIAL218790-50G SIAL218790-5G
Tetraphenylphosphonium chloride, Sigma-Aldrich®
Tetraphenylphosphonium chloride
Formula: C₂₄H₂₀ClP
MW: 374.85 g/mol
Melting Pt: 275…278 °C
MDL Number: MFCD00011916
CAS Number: 2001-45-8
EINECS: 217-890-3