Ammonium chloride, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
MQ200
326372-100G
326372-25G
326372-500G
SIAL326372-100GEA
193
EUR
SIAL326372-100G
SIAL326372-25G
SIAL326372-500G
Ammonium chloride, Sigma-Aldrich®
Ammonium chloride
Formula:
NH₄Cl MW: 53.49 g/mol Melting Pt: 335 Density: 1.5256 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...