Ammonium nitrate, Sigma-Aldrich®

Supplier: Merck
Warning


MQ200

A7455-2.5KG A7455-500G
SIALA7455-2.5KGEA 270 EUR
SIALA7455-2.5KG SIALA7455-500G
Ammonium nitrate, Sigma-Aldrich®
Ammonium nitrate
Formula: NH₄NO₃
MW: 80.04 g/mol
Boiling Pt: 230
Melting Pt: 169
Density: 1.73 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1 III
Merck Index: 13,00538

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR