Ammonium sulphate
Supplier: Roth Carl
Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
3746.1 3746.2 3746.3 3746.4 3746.5
ROTH3746.1EA 0 EUR
ROTH3746.1 ROTH3746.2 ROTH3746.3 ROTH3746.4 ROTH3746.5
Ammonium sulphate
Ammonium sulphate
Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 336…339 °C
Density: 1.78 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1
REACH: 01-2119455044-46
Merck Index: 13,00559