Ammonium iron(II) sulphate
Supplier: Lovibond
LOVI460111EA 63.4 EUR
LOVI460111
Ammonium iron(II) sulphate
Ammonium iron(II) sulphate
Formula: H₈FeN₂O₈S₂
MW: 284,05 g/mol
Melting Pt: 100 °C
Density: 1,86 g/cm³ (20 °C)
MDL Number: MFCD00051039
UN: 2796
ADR: 8,II
Merck Index: 12,00552