Thionyl bromide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Thionyl dibromide
M-Clarity™ quality level = MQ200
251259-100G 251259-25G
SIAL251259-100GEA 176 EUR
SIAL251259-100G SIAL251259-25G
Thionyl bromide, Sigma-Aldrich®
Thionyl bromide
Formula: Br₂OS
Boiling Pt: 47…49 °C (20 mmHg)
Melting Pt: –49.5 °C
Density: 2.672 g/cm³ (25 °C)
MDL Number: MFCD00011448
CAS Number: 507-16-4
EINECS: 208-064-3
UN: 3264
ADR: 8,I
Merck Index: 12,09484