Disulphur dichloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Sulphur monochloride
M-Clarity™ quality level = MQ200
157759-1KG 157759-250G 157759-50G
SIAL157759-1KGEA 66.4 EUR
SIAL157759-1KG SIAL157759-250G SIAL157759-50G
Disulphur dichloride, Sigma-Aldrich®
Disulphur dichloride
Formula: Cl₂S₂
MW: 135.04 g/mol
Boiling Pt: 138 °C (1013 hPa)
Melting Pt: –76 °C
Density: 1.68 g/cm³ (20 °C)
Flash Pt: 118 °C
Storage Temperature: Ambient
CAS Number: 10025-67-9
EINECS: 233-036-2
UN: 1828
ADR: 8,I
Merck Index: 12,09143