Selenium tetrachloride, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: Selenium(IV) chloride

M-Clarity™ quality level = MQ100

323527-10G 323527-50G
SIAL323527-10GEA 70.2 EUR
SIAL323527-10G SIAL323527-50G
Selenium tetrachloride, Sigma-Aldrich®
Selenium tetrachloride
Formula: Cl₄Se
MW: 220.77 g/mol
Melting Pt: 330 °C
Density: 2.68 g/cm³ (20 °C)
MDL Number: MFCD00011225
CAS Number: 10026-03-6
EINECS: 233-053-5
UN: 2923
ADR: 8,II
Merck Index: 12,08582

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR