Tellurium tetrachloride, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Tellurium(IV) chloride
M-Clarity™ quality level = MQ100
205338-100G 205338-25G 205338-5G
SIAL205338-100GEA 719 EUR
SIAL205338-100G SIAL205338-25G SIAL205338-5G
Tellurium tetrachloride, Sigma-Aldrich®
Tellurium tetrachloride
Formula: Cl₄Te
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 224 °C
Density: 3.01 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011262
CAS Number: 10026-07-0
EINECS: 233-055-6
UN: 3260
ADR: 8,II
Merck Index: 12,09279