Chlorosulphuric acid 97%
Supplier: Thermo Scientific
Danger
Synonyms: Chlorosulphonic acid
304490010 304491000 304495000 304490025
ACRO304490010EA 82.1 EUR
ACRO304490010 ACRO304491000 ACRO304495000 ACRO304490025
Chlorosulphuric acid 97%
Chlorosulphuric acid
Formula: HClO₃S
MW: 116.52 g/mol
Boiling Pt: 151…152 °C (760 mmHg)
Melting Pt: –80 °C
Density: 1.75 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8,
Merck Index: 12,02218
Order Now
Specification Test Results
Appearance Slightly hazy viscous liquid
Titration ≥96.0 %