Chlorosulphuric acid, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Chlorosulphonic acid
M-Clarity™ quality level = MQ200
571024-100G
571024-1KG
571024-5G
SIAL571024-100GEA
104
EUR
SIAL571024-100G
SIAL571024-1KG
SIAL571024-5G
Chlorosulphuric acid, Sigma-Aldrich®
Chlorosulphuric acid
Formula:
HClO₃S MW: 116.52 g/mol Boiling Pt: 152 °C (1013 hPa) Melting Pt: –80 °C Density: 1.75 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011523 CAS Number: 7790-94-5 EINECS: 232-234-6 UN: 1754 ADR: 8,I Merck Index: 12,02218 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...