Amidosulphonic acid
Supplier: Hach
HACH105599EA 37.3 EUR
HACH105599
Amidosulphonic acid
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97,09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2,15 g/cm³ (20 °C)
MDL Number: MFCD00011603
UN: 2967
ADR: 8,III
Merck Index: 13,09006
https://si.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg