Amidosulphonic acid

Supplier: Hach

HACH2076266EA 62.9 EUR
HACH2076266
Amidosulphonic acid
Amidosulphonic acid
Formula: H₃NO₃S
MW: 97,09 g/mol
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2,15 g/cm³ (20 °C)
MDL Number: MFCD00011603
UN: 2967
ADR: 8,III
Merck Index: 13,09006
https://si.vwr.com/stibo/web/std.lang.all/75/87/2297587.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR