Amidosulphonic acid ≥99%, TECHNICAL
Supplier: VWR Chemicals
Warning
Synonyms: Aminosulphonic acid, Amidosulphuric acid, Amidosulphuric Acid, Sulphamic acid, Amidosulphonic acid, Sulphamidic acid
20671.297EA 46.1 EUR
20671.297 20671.468 20671.366
Amidosulphonic acid ≥99%, TECHNICAL
Amidosulphonic acid
Formula: H₃NO₃S
Boiling Pt: 247 °C (1013 hPa)
Melting Pt: 205 °C
Density: 2.15 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011603
CAS Number: 5329-14-6
EINECS: 226-218-8
UN: 2967
ADR: 8,III
Merck Index: 13,09006
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afa0500786be69d531cdf06f8e67548e6f67b23f&vid=73aa20196fbabf965e4375f2cf3675e2f513ee59&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay Min. 99 %