Disulphiram, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Tetraethylthiuram disulphide, 1,1',1'',1'''-{Disulphanediylbis[(thioxomethylene)-nitrilo]}tetraethane, Bis(diethylthiocarbamoyl) disulphide
86720-1KG 86720-250G 86720-50G
SIAL86720-1KGEA 120 EUR
SIAL86720-1KG SIAL86720-250G SIAL86720-50G
Disulphiram, Sigma-Aldrich®
Disulphiram
Formula: C₁₀H₂₀N₂S₄
MW: 296.55 g/mol
Boiling Pt: 369 °C (1013 hPa)
Melting Pt: 70 °C
Density: 1.34 g/cm³ (20 °C)
MDL Number: MFCD00009048
CAS Number: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 13,03399