Dimethyl sulphoxide 99.8+%, extra pure
Supplier: Thermo Fisher Scientific
Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
295520010 295522500 295520025 295520250
ACRO295520010EA 80.5 EUR
ACRO295520010 ACRO295522500 ACRO295520025 ACRO295520250
Dimethyl sulphoxide 99.8+%, extra pure
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (760 mmHg)
Melting Pt: 18.4 °C
Density: 1.1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Clear liquid
Water ≤0.1 %
GC ≥99.80 %
Color scale ≤10 APHA
Residue after evaporation ≤0.003 %
Acidity ≤0.04 mg KOH/g