Dimethyl sulphoxide, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ500 (more info.)
D2438-50ML
D2438-5X10ML
SIALD2438-50MLEA
183
EUR
SIALD2438-50ML
SIALD2438-5X10ML
Dimethyl sulphoxide, Sigma-Aldrich®
Dimethyl sulphoxide
Formula:
(CH₃)₂SO Boiling Pt: 189 °C (1013 hPa) Melting Pt: 18.5 °C Density: 1.101 g/cm³ (20 °C) Flash Pt: 87 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...