Dimethyl sulphoxide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO
M-Clarity™ quality level = MQ200
D8418-100ML
D8418-250ML
D8418-50ML
SIALD8418-100MLEA
114
EUR
SIALD8418-100ML
SIALD8418-250ML
SIALD8418-50ML
Dimethyl sulphoxide, Sigma-Aldrich®
Dimethyl sulphoxide
Formula:
(CH₃)₂SO MW: 78.14 g/mol Boiling Pt: 189 Melting Pt: 18.5 Density: 1.101 g/cm³ (20 °C) Flash Pt: 87 Storage Temperature: Ambient |
MDL Number:
MFCD00002089 CAS Number: 67-68-5 EINECS: 200-664-3 Merck Index: 13,03285 |