Dibutyl disulphide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Dibutyldisulphide, n-Butyl disulphide
M-Clarity™ quality level = MQ100
B93989-100ML B93989-25ML
SIALB93989-100MLEA 126 EUR
SIALB93989-100ML SIALB93989-25ML
Dibutyl disulphide, Sigma-Aldrich®
Dibutyl disulphide
Formula: C₈H₁₈S₂
MW: 178.36 g/mol
Boiling Pt: 229
Melting Pt: –94 °C
Density: 0.9371 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009467
CAS Number: 629-45-8
EINECS: 211-091-3