Diethyl disulphide, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Ethyldisulphide
M-Clarity™ quality level = MQ300
W409301-SAMPLE W409301-100G W409301-1KG
SIALW409301-SAMPLEEA 54.3 EUR
SIALW409301-SAMPLE SIALW409301-100G SIALW409301-1KG
Diethyl disulphide, Sigma-Aldrich®
Diethyl disulphide
Formula: C₄H₁₀S₂
MW: 122.26 g/mol
Boiling Pt: 152 °C (1013 hPa)
Melting Pt: –101.5 °C
Density: 0.984 g/cm³ (20 °C)
MDL Number: MFCD00009266
CAS Number: 110-81-6
EINECS: 203-805-7
UN: 1993
ADR: 3,III