Dimethyl diselenide, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
Synonyms:
Dimethyldiselenide
M-Clarity™ quality level = MQ100
328502-1G
328502-5G
SIAL328502-1GEA
73.3
EUR
SIAL328502-1G
SIAL328502-5G
Dimethyl diselenide, Sigma-Aldrich®
Dimethyl diselenide
Formula:
C₂H₆Se₂ MW: 187.99 g/mol Boiling Pt: 155 Melting Pt: –82 °C Density: 2.0009 g/cm³ (20 °C) |
MDL Number:
MFCD00008254 CAS Number: 7101-31-7 EINECS: 230-410-7 UN: 2929 ADR: 6.1 II |