Dimethyl sulphide, Sigma-Aldrich®
Supplier: Merck
Danger
471577-250ML 471577-25ML
SIAL471577-250MLEA 65.7 EUR
SIAL471577-250ML SIAL471577-25ML
Dimethyl sulphide, Sigma-Aldrich®
Dimethyl sulphide
Formula: C₂H₆S
Boiling Pt: 36.2 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.85 g/cm³ (20 °C)
Flash Pt: –18 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008562
CAS Number: 75-18-3
EINECS: 200-846-2
UN: 1164
ADR: 3,II
Merck Index: 12,06204