Dimethyl trisulphide, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Dimethyltrisulphide
M-Clarity™ quality level = MQ300
W327506-100G-K
W327506-1KG-K
SIALW327506-100G-KEA
336
EUR
SIALW327506-100G-K
SIALW327506-1KG-K
Dimethyl trisulphide, Sigma-Aldrich®
Dimethyl trisulphide
Formula:
C₂H₆S₃ MW: 126.27 g/mol |
MDL Number:
MFCD00039808 CAS Number: 3658-80-8 EINECS: 222-910-9 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...