Dipropyl disulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms:
1,2-Dipropyldisulphane, Propyl disulphide
M-Clarity™ quality level = MQ300
W322806-100G-K
W322806-1KG-K
W322806-5KG-K
SIALW322806-100G-KEA
78.6
EUR
SIALW322806-100G-K
SIALW322806-1KG-K
SIALW322806-5KG-K
Dipropyl disulphide, Sigma-Aldrich®
Dipropyl disulphide
Formula:
C₆H₁₄S₂ MW: 150.31 g/mol Boiling Pt: 195 °C (1013 hPa) Melting Pt: –85.6 °C Density: 0.9598 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009378 CAS Number: 629-19-6 EINECS: 211-079-8 UN: 3082 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...