Ethyl methyl sulphide, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

238317-100G 238317-25G
SIAL238317-100GEA 138 EUR
SIAL238317-100G SIAL238317-25G
Ethyl methyl sulphide, Sigma-Aldrich®
Ethyl methyl sulphide
Formula: C₃H₈S
MW: 76.16 g/mol
Boiling Pt: 65
Melting Pt: –104.8 °C
Density: 0.837 g/cm³ (20 °C)
MDL Number: MFCD00009268
CAS Number: 624-89-5
EINECS: 210-868-4
UN: 1993
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR