2,2'-Dipyridyl disulphide, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,2-Di(pyridin-2-yl)disulphane, 2,2'-Dithiodipyridine
M-Clarity™ quality level = MQ100
D5767-1G D5767-25G D5767-5G
SIALD5767-1GEA 36.9 EUR
SIALD5767-1G SIALD5767-25G SIALD5767-5G
2,2'-Dipyridyl disulphide, Sigma-Aldrich®
2,2'-Dipyridyl disulphide
Formula: C₁₀H₈N₂S₂
MW: 220.32 g/mol
Boiling Pt: 348
Melting Pt: 57…61 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006287
CAS Number: 2127-03-9
EINECS: 218-343-1