Phenylhydrazine, Sigma-Aldrich®

Supplier: Merck

P26252-100G P26252-500G P26252-5G
SIALP26252-100GEA 51.1 EUR
SIALP26252-100G SIALP26252-500G SIALP26252-5G
Phenylhydrazine, Sigma-Aldrich®
Phenylhydrazine
Formula: C₆H₈N₂
MW: 108.14 g/mol
MDL Number: MFCD00007573
CAS Number: 100-63-0
Merck Index: 12,07447

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR