1-Methylimidazole (NMI), Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

M-Clarity™ quality level = MQ300

336092-100ML 336092-1L 336092-2L
SIAL336092-100MLEA 115 EUR
SIAL336092-100ML SIAL336092-1L SIAL336092-2L
1-Methylimidazole (NMI), Sigma-Aldrich®
1-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 3267
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR