1-Methylimidazole (NMI), Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
M50834-100G M50834-500G
SIALM50834-100GEA 42.1 EUR
SIALM50834-100G SIALM50834-500G
1-Methylimidazole (NMI), Sigma-Aldrich®
1-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.03 g/cm³ (20 °C)
Flash Pt: 92 °C
Storage Temperature: Ambient
MDL Number: MFCD00005292
CAS Number: 616-47-7
EINECS: 210-484-7
UN: 3267
ADR: 8,II