2-Methylimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Methyl-1H-imidazole
M-Clarity™ quality level = MQ200
M50850-100G M50850-500G
SIALM50850-100GEA 54 EUR
SIALM50850-100G SIALM50850-500G
2-Methylimidazole, Sigma-Aldrich®
2-Methylimidazole
Formula: C₄H₆N₂
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 130…133 °C
Density: 0.99 g/cm³ (25 °C)
Flash Pt: 155 °C
Storage Temperature: Ambient
MDL Number: MFCD00005190
CAS Number: 693-98-1
EINECS: 211-765-7
UN: 3259
ADR: 8,III