1-Benzylimidazole 98
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 1-Benzyl-1H-imidazole
L00802.06 L00802.14
L00802.06EA 38.2 EUR
L00802.06 L00802.14
1-Benzylimidazole 98
1-Benzylimidazole
Purity: 98+% CAS No.: 4238-71-5 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 68-73° Boiling Point: 310° MDL No.: MFCD00005296
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 310 °C
Melting Pt: 68…73 °C
Density: 1.22 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005296
CAS Number: 4238-71-5
EINECS: 224-200-4