1-Benzylimidazole
Supplier: Cayman Chemical
Synonyms: 1-Benzyl-1H-imidazole
70510-5 70510-25 70510-10
CAYM70510-5EA 48 EUR
CAYM70510-5 CAYM70510-25 CAYM70510-10
1-Benzylimidazole
1-Benzylimidazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 310
Melting Pt: 72
Density: 1.22 g/cm³ (25 °C)
MDL Number: MFCD00005296
CAS Number: 4238-71-5
EINECS: 224-200-4