1-Benzylimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Benzyl-1H-imidazole
MQ100
116416-25G 116416-5G
SIAL116416-25GEA 71.3 EUR
SIAL116416-25G SIAL116416-5G
1-Benzylimidazole, Sigma-Aldrich®
1-Benzylimidazole
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 310
Melting Pt: 72
Density: 1.22 g/cm³ (25 °C)
MDL Number: MFCD00005296
CAS Number: 4238-71-5
EINECS: 224-200-4