1,1'-Thiocarbonyldiimidazole, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1'-Thiocarbonylbis(imidazole), 1,1'-Thiocarbonylbis-1H-imidazole
88545-1G 88545-25G 88545-5G
SIAL88545-1GEA 41.6 EUR
SIAL88545-1G SIAL88545-25G SIAL88545-5G
1,1'-Thiocarbonyldiimidazole, Sigma-Aldrich®
1,1'-Thiocarbonyldiimidazole
Formula: C₇H₆N₄S
Melting Pt: 105…106 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005289
CAS Number: 6160-65-2
EINECS: 228-183-4