1-Indanone, Sigma-Aldrich®
Supplier: Merck
Synonyms: 2,3-Dihydro-1H-inden-1-one
M-Clarity™ quality level = MQ100
I2304-100G I2304-10G I2304-25G
SIALI2304-100GEA 327 EUR
SIALI2304-100G SIALI2304-10G SIALI2304-25G
1-Indanone, Sigma-Aldrich®
1-Indanone
Formula: C₉H₈O
MW: 132.16 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 37…40 °C
Density: 1.1028 g/cm³ (40 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003785
CAS Number: 83-33-0
EINECS: 201-470-1